Furnace Cap Hood, Monashee
Supplier: Monashee
107239-05 107239-05
11022-166EA 2355.1 USD
CA11022-166 11022-166
Furnace Cap Hood, Monashee
Fume Hoods and Enclosures Ductless Fume Hoods
Furnace cap hoods are designed to capture the lead fumes from your MAS3 fusion or cupellation furnaces. Designed for fire assay applications, our furnace cap hoods have a 10" flange for connecting to existing duct work and are required to be connected to an appropriately sized dust collector and a scrubber as required (items sold separately, contact us for more information).
  • Designed to fit over the MAS 2 and MAS 3 furnaces
  • Solid steel construction with powder coating for long lasting finish
  • 10" flange to attach to ducting
Furnace cap hoods are made of steel and white powder coated for a long-lasting finish (additional colors available on request). Furnace cap hoods for other models of furnaces are also available upon request.