Adafruit N20 DC Gearmotor with Magnetic Encoder, Digikey
Supplier: DIGI-KEY 465986 MS
1528-4638-ND 1528-4639-ND 1528-4640-ND 1528-4641-ND
MSPP-1528-4638EA 19.23 USD
MSPP-1528-4638 MSPP-1528-4639 MSPP-15284640N MSPP-15284641N
Adafruit N20 DC Gearmotor with Magnetic Encoder, Digikey
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
This N20 DC motor comes with magnetic encoder. As the motor turns, the attached encoder wheel spins, causing the counter to detect each passing spoke and that lets your microcontroller count and determine speed.
  • Varied rpm
  • Comes in bulk package
  • Has magnetic wheel and two hall effect sensors already attached
  • Power 0.2 W
Provide 4.5 to 6 V DC (nominal) to the white and red wires - these connect to your motor driver, and can be PWM'd for speed adjustment and direction by using an H-bridge.
Certifications: RoHS compliant.