Huma OsteoGelMA High Bloom Methacrylated Gelatin, Lyophilized, Humabiologics
Supplier: Humabiologics
BMHL-1G
76452-640EA 477.79 USD
76452-640
Huma OsteoGelMA High Bloom Methacrylated Gelatin, Lyophilized, Humabiologics
Cell Culture Supplements and Additives
Clinically relevant native human methacrylated bone gelatin for hydrogel and bioink applications.
  • Native human methacrylated bone gelatin
  • Clinically relevant and xeno-free for regenerative medicine applications
  • Lyophilized for tailored 3D hydrogel or bioink applications
  • Supports various cell attachments
Huma OsteoGelMA is isolated from human bone sourced strictly from tissue partners who comply with the requirements for transplantable human tissue. Huma OsteoGelMA has undergone a proprietary manufacturing method to extract and methacrylate human gelatin from bone. Huma OsteoGelMA bone-derived gelatin is supplied lyophilized for hydrogel, bioink, or cell culture applications.
Huma OsteoGelMA addresses the growing need for more clinically relevant and animal-free human tissue constructs and models. Each lot of Huma OsteoGelMA has undergone extensive testing to meet high safety and quality requirements for translational research needs.
The safety of all tissue has been tested and found negative for communicable diseases and infectious agents.
Ordering information: Huma OsteoGelMA High Bloom is available in 1 g.
Caution: Huma OsteoGelMA is for research use only and is not intended for human use, diagnosis, screening, household, food or other uses.