Sodium chloride solution 0,45% USP for injection

Supplier: Fresenius Kabi

Synonyms: Saline, NaCl, Sea salt, Halite

63323-626-50 63323-626-00 63323-626-25 63323-626-55 63323-626-10
76460-908CS 362.16 USD
76460-908 76460-910 76460-912 76460-914 76460-916
Sodium chloride solution 0,45% USP for injection
Sodium chloride

  • Preservative Free
  • Bar Coded
  • AP Rated
  • Non-DEHP
  • Non-PVC

Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Merck Index: 14,08599

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR