HelixMark® Special Platinum-Cured Tubing, Wacker Material, Freudenberg Medical
Supplier: Freudenberg Medical
HelixMark®
61-747-194 61-747-243 61-747-98
32828-034EA 3942.72 USD
32828-034 32828-012 32828-032
HelixMark® Special Platinum-Cured Tubing, Wacker Material, Freudenberg Medical
Tubing Transfer Tubing
HelixMark® platinum-cured silicone tubing meets the needs of a broad range of applications and is universally applicable for the transfer of fluids under standard requirements.
  • Fully autoclavable and sterilizable
  • Ideal for use in transfer operations as well as other applications requiring non-leachable material
  • Material certification in every package
HelixMark® tubing meets USP Class VI and EP 3.1.9 biocompatiblity requirements. It is manufactured in Freudenberg Medical's FDA registered medical device facility following Good Manufacturing Practices (GMPs) in accordance with The Quality Systems Requlations.
Certified to meet ISO 13485 quality standards.
Durometer hardness: Shore A, 60.
Certifications: Materials approved in accordance with USP Class VI, ISO 10993-1, FDA 21CFR 177-2600 and EP 3.1.9 or EP 3.1.9 without appearance of solution.
Packaging: Supplied with 5×50 ft. coils per box. Each coil is double bagged.