QC Sample pH 7, Color Coded Yellow, SPEX CertiPrep
Supplier: SPEX CertiPrep
132 131
SX131EA 31.44 USD
SX131 SX132
QC Sample pH 7, Color Coded Yellow, SPEX CertiPrep
Buffers pH Calibration Buffers
QC sample directly traceable to NIST SRMs, mercury free, guaranteed stable for at least one year after your receipt, and supplied with a full certificate of analysis
The end-product of most analytical laboratories is data. It is therefore imperative that the quality of the data produced meet or exceed performance criteria. The implementation of Quality Control (QC) processes helps control error and estimate uncertainty. QC can be a tool in method development, regulatory processes, laboratory training, and performance monitoring. QC samples can help an analytical laboratory determine its precision, accuracy, linear range, and methof ruggedness.
QC samples allow the laboratories to fulfill documentation requirements as well as correct errors in root case analysis of a possible failure. Finally, QC samples monitor the effectiveness of the laboratory analysts to produce accurate data.