Nitric acid 65.0 - 71.0%, CMOS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Nitric acid solution
Management of Change (MOC) category = R
9685-69
BAKR9685-69
JT9685-69EA
3.77
USD
CAJT9685-69
JT9685-69
Nitric acid 65.0 - 71.0%, CMOS, J.T.Baker®
Nitric acid
Formula:
HNO₃ MW: 63.01 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011349 CAS Number: 7697-37-2 UN: 2031 ADR: 8,II |
Specification Test Results
For Microelectronic Use | |
Assay (HNO₃) | 65.0 - 71.0 % |
Color, APHA | ≤10 |
Chloride (Cl) | ≤80 ppb |
Sulfate (SO4) | ≤500 ppb |
Phosphate (PO₄) | ≤200 ppb |
Arsenic (As) | ≤100.0 ppb |
Copper (Cu) | ≤100.0 ppb |
Lead (Pb) | ≤100.0 ppb |
Iron (Fe) | ≤100.0 ppb |
Sodium (Na) | ≤100.0 ppb |
Boron (B) | ≤100.0 ppb |
Magnesium (Mg) | ≤100.0 ppb |
Aluminum (Al) | ≤100.0 ppb |
Potassium (K) | ≤100.0 ppb |
Calcium (Ca) | ≤100.0 ppb |
Chromium (Cr) | ≤100.0 ppb |
Nickel (Ni) | ≤100.0 ppb |
Zinc (Zn) | ≤100.0 ppb |
Silver (Ag) | ≤100.0 ppb |
Barium (Ba) | ≤100.0 ppb |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...