Add-Ons Mix for US EPA Methods 625 and 8270, SPEX CertiPrep
Supplier: SPEX CertiPrep
ECS-N-031 ECS-B-031 ECS-A-031
SXECS-N-031EA 142 USD
SXECS-N-031 SXECS-A-031 SXECS-B-031
Add-Ons Mix for US EPA Methods 625 and 8270, SPEX CertiPrep
Standards Environmental Analysis Standards
ECS Standards for GC/MS
All ampulized standards are supplied with a pre-labeled transfer/storage vial. We want to make the transfer of ampulized standards as easy as possible. The label includes the catalog number, lot number, product description and expiration date.