Pulverizer Hoods, Monashee
Supplier: MONASHEE MANUFACTURING CA
107239-03L 107239-03R
CA11022-158EA 5151.55 USD
CA11022-158 CA11022-160
Pulverizer Hoods, Monashee
Fume Hoods and Enclosures Ductless Fume Hoods
Pulverizer hoods effectively capture and collect dust while providing uniform airflow, a clean work surface and protecting the operator from contaminants. Hood can have the option to have your pulverizer placed on the left- or right-hand side to provide a convenient work area for your pulverizer bowls, discs, and sample preparation. Designed for mining applications, our workstation dust hoods have a 10" flange for connecting to existing duct work and are required to be connected to an appropriately sized dust collector (sold separately, contact us for more information).
  • Solid steel
More Product Information