Avantor® Hichrom HI-5 MS Xil, GC Columns
Supplier: VWR
Avantor® Hichrom
HI19-53-025-30 HI19-20-020-60 HI19-53-100-30 HI19-18-020-20 HI192502560G5 HI19-53-100-10 HI19-18-018-20 HI19-20-040-40 HI193202530G5 HI19-25-050-50 HI19-53-150-15 HI19-20-010-50 HI19-53-050-10 HI19-20-040-20 HI19-53-100-50 HI19-32-010-15 HI19-18-020-50 HI19-32-100-15 HI19-18-036-20 HI19-10-020-5 HI19-32-050-15 HI192505030G5 HI19-15-015-30 HI19-18-040-60 HI19-32-150-30 HI1925025105 HI19-25-050-15 HI19-32-010-30 HI19-18-040-20 HI1932010105 HI1925100105 HI19-32-010-50 HI19-53-150-60 HI19-10-040-10 HI19-20-010-40 HI19-53-025-60 HI19-53-025-50 HI19-32-100-10 HI192505015G10 HI19-20-020-50 HI19-20-040-10 HI19-32-150-60 HI192505030G10 HI195310030G5 HI19-32-100-50 HI19-20-040-60 HI19-20-033-50 HI19-10-020-15 HI19-32-150-15 HI19-32-025-60 HI19-32-100-60 HI1932100105 HI19-20-010-60 HI19-18-040-10 HI19-10-010-5 HI19-32-100-30 HI19-25-100-60 HI19-20-040-50 HI19-20-020-20 HI19050052.5 HI19-20-033-60 HI19-25-025-30 HI19-32-025-50 HI19-32-050-60 HI19-32-010-10 HI19-53-150-75 HI19-53-010-15 HI19-20-033-12 HI19-20-100-60 HI19-20-020-10 HI19-18-018-60 HI192501030G5 HI19-25-010-60 HI19-53-050-30 HI19-53-150-10 HI1925050105 HI19-20-100-50 HI19-53-025-15 HI19-20-010-10 HI19-32-050-50 HI19-18-018-40 HI19-10-020-10 HI19-18-036-40 HI19-05-005-5 HI19-25-025-10 HI19-15-015-15 HI19-20-100-20 HI19-10-040-5 HI19-25-010-30 HI19-25-050-30 HI19-18-020-60 HI1932025105 HI19-25-100-10 HI19-15-015-20 HI192502530G5 HI19-15-015-10 HI19-10-010-15 HI192502560G10 HI19-18-020-10 HI19-18-040-50 HI19-32-025-30 HI19-25-100-30 HI19-25-025-60 HI19-18-018-10 HI19-25-025-15 HI19-32-050-10 HI192502530G10 HI19-53-010-10 HI19-20-033-30 HI19-20-100-40 HI19-25-025-50 HI19-32-150-10 HI19-25-050-10 HI19-53-100-75 HI19-18-018-50 HI19-25-050-60 HI19-53-150-50 HI19-32-025-10 HI19-25-100-15 HI19-53-050-15 HI19-53-100-60 HI19-20-020-40 HI19-25-010-15 HI19-25-100-50 HI19-20-100-10 HI19-32-050-30 HI19-32-025-15 HI1932150105 HI19-53-050-50 HI19-32-010-60 HI19-20-010-20 HI192502515G5 HI1932050105 HI19-20-033-25 HI19-53-010-30 HI19-32-150-50 HI19-53-050-60 HI19-53-100-15 HI19-25-010-50 HI19-05-005-2 HI19-10-010-10 HI19-18-036-60 HI19-15-015-5 HI19-18-020-40 HI19-18-040-40 HI19-53-150-30 HI193210030G5 HI19-10-040-15
76434-060EA 1007.03 USD
76434-060 76434-062 76434-064 76434-066 76434-068 76431-254 76431-252 76433-262 76433-142 76431-250 76433-260 76433-140 76433-266 76433-146 76433-300 76433-264 76433-144 76431-258 76433-304 76433-268 76433-148 76433-302 76431-256 76434-050 76434-052 76434-054 76434-056 76434-058 76433-130 76433-252 76433-250 76433-134 76433-256 76433-132 76433-254 76433-138 76433-136 76433-258 76433-118 76434-044 76434-046 76434-048 76433-240 76433-120 76433-244 76433-124 76433-242 76433-122 76431-238 76433-248 76431-236 76433-128 76433-246 76433-126 76433-228 76433-192 76431-242 76431-240 76433-190 76433-350 76433-196 76433-230 76433-194 76433-234 76431-248 76433-198 76433-232 76431-246 76433-116 76433-238 76431-244 76433-114 76433-236 76433-338 76433-218 76433-182 76433-180 76433-186 76433-340 76433-184 76433-222 76433-344 76433-220 76433-342 76433-188 76433-226 76433-348 76433-224 76433-346 76433-208 76433-206 76433-328 76433-170 76433-292 76433-290 76433-174 76433-296 76433-172 76433-294 76433-332 76433-178 76433-212 76433-176 76433-330 76433-298 76433-210 76433-336 76433-216 76433-334 76433-214 76433-318 76433-316 76433-280 76433-160 76433-284 76433-164 76433-282 76433-162 76433-288 76433-200 76433-168 76433-322 76433-286 76433-166 76433-320 76433-204 76433-326 76433-202 76433-324 76433-308 76434-070 76433-306 76434-072 76434-074 76434-076 76433-270 76433-152 76433-274 76433-150 76433-272 76433-156 76433-310 76433-278 76433-154 76433-276 76433-314 76433-312 76433-158
Avantor® Hichrom HI-5 MS Xil, GC Columns
Chromatography Columns
Silphenylene based stationary phase (selectivity equivalent to 5MS columns) to ensure minimum bleeding level at high temperatures. General purpose low-polarity phase for a wide range of applications, including (for example): PCBs, essential oils, pesticides, semivolatiles and many more. HI-5 MS Xil is an MS grade low-bleeding crossbonded column and assure a long lifetime even under high temperature limits.
  • Phase composition: Crossbonded Silphenylene - Methyl Polysiloxane (5MS column selectivity) - MS grade
  • Long lifetime
Max temperature limits may change depending on the film thickness.