Avantor® Hichrom HI-1, GC Columns
Supplier: VWR
Avantor® Hichrom
HI01-20-010-40 HI01-18-018-60 HI01-32-400-30 HI0125050105 HI01-53-500-50 HI015310030G5 HI01-53-025-50 HI01-53-010-10 HI01-25-025-60 HI01-32-050-10 HI01-20-040-40 HI01-53-010-30 HI01-20-100-20 HI0132010105 HI01-25-050-60 HI01-53-265-10 HI01-25-010-30 HI01-32-050-15 HI01-53-100-15 HI01-18-040-60 HI01-20-100-60 HI01-53-300-10 HI01-10-040-5 HI01-53-300-60 HI01-20-040-20 HI01-53-100-50 HI01-25-050-50 HI01-25-025-15 HI01-32-500-30 HI01-53-300-50 HI01-20-020-60 HI01-18-040-20 HI01-53-150-15 HI01-32-300-30 HI01-32-300-15 HI01-32-150-50 HI01-53-150-10 HI01-10-010-15 HI01-32-010-15 HI01-53-500-75 HI01-15-015-10 HI01-25-050-10 HI01-32-100-15 HI01-20-020-40 HI01-53-500-10 HI01-25-100-60 HI01-18-018-40 HI01-25-025-10 HI01-20-010-60 HI01-18-018-50 HI01-15-015-5 HI01-18-018-20 HI01-53-010-15 HI01-32-100-30 HI0132025105 HI0132500105 HI01-10-020-10 HI01-10-010-5 HI01-53-050-30 HI01-32-100-60 HI01-32-010-60 HI01-25-100-30 HI01-10-040-10 HI01-32-050-50 HI01-25-100-10 HI01-32-150-15 HI01-15-015-20 HI01-25-025-30 HI01-53-500-30 HI012502560G5 HI01-20-040-10 HI01-20-020-10 HI01-18-040-40 HI01-18-020-10 HI01-32-010-10 HI0132100105 HI01050052.5 HI01-32-500-10 HI01-20-040-50 HI01-15-015-15 HI01-53-150-50 HI01-53-025-15 HI01-20-020-50 HI01-32-025-60 HI01-32-025-10 HI01-05-005-2 HI01-53-300-75 HI01-53-050-50 HI01-32-025-30 HI01-18-040-10 HI01-53-100-75 HI01-53-500-60 HI01-20-020-20 HI01-32-150-10 HI012502515G5 HI01-18-020-40 HI0125025105 HI01-18-020-20 HI01-53-700-15 HI01-25-010-60 HI01-25-010-15 HI01-53-300-30 HI01-32-150-60 HI01-20-040-60 HI01-25-050-30 HI01-20-010-50 HI01-15-015-30 HI01-53-500-15 HI01-25-100-50 HI01-32-050-60 HI01-32-010-30 HI01-20-100-40 HI01-53-300-15 HI01-25-100-15 HI01-32-025-50 HI01-32-010-50 HI01-18-040-50 HI01-53-050-15 HI01-32-100-10 HI012502530G5 HI01-32-500-15 HI0132300105 HI01-32-050-30 HI01-20-100-10 HI01-53-050-10 HI01-18-018-10 HI01-05-005-5 HI01-10-040-15 HI01-32-300-60 HI01-32-100-50 HI01-53-150-30 HI01-53-150-60 HI01-10-020-5 HI01-10-010-10 HI01-32-150-30 HI01-32-300-10 HI01-53-050-60 HI01-53-100-10 HI01-10-020-15 HI0125100105 HI01-20-010-10 HI01-53-700-30 HI01-25-010-50 HI01-53-100-60 HI0132150105 HI01-32-500-50 HI0132050105 HI01-20-100-50 HI01-25-025-50 HI01-53-150-75 HI01-53-025-60 HI01-53-025-30 HI01-32-025-15 HI01-25-050-15 HI01-53-100-30 HI01-20-010-20 HI01-32-500-60 HI01-32-300-50
76434-180EA 835.77 USD
76434-180 76434-182 76434-184 76434-340 76434-186 76434-220 76434-188 76434-342 76434-222 76434-344 76434-224 76434-346 76434-226 76434-216 76434-338 76434-218 76434-290 76434-170 76434-292 76434-172 76434-294 76434-174 76434-296 76434-330 76434-176 76434-298 76434-210 76434-178 76434-332 76434-212 76434-334 76434-214 76434-336 76434-326 76434-206 76434-328 76434-208 76430-102 76430-104 76434-280 76434-160 76430-106 76434-282 76434-162 76430-108 76434-284 76434-164 76434-286 76434-320 76434-166 76434-288 76434-200 76434-168 76434-322 76434-202 76430-100 76434-324 76434-204 76434-316 76434-318 76434-390 76434-270 76434-392 76434-272 76434-394 76434-274 76434-154 76434-396 76434-276 76434-398 76434-156 76434-310 76434-278 76434-158 76434-312 76434-314 76434-304 76434-426 76434-306 76434-308 76430-124 76430-126 76434-380 76434-260 76434-382 76434-262 76434-384 76434-264 76434-386 76434-266 76434-420 76434-388 76430-120 76434-300 76434-268 76434-422 76430-122 76434-302 76434-424 76434-414 76431-056 76434-416 76431-054 76434-418 76431-052 76431-050 76430-114 76430-116 76430-118 76434-370 76434-250 76434-372 76434-252 76434-374 76434-254 76434-376 76434-410 76434-256 76430-110 76434-378 76434-412 76434-258 76430-112 76434-404 76434-406 76434-408 76434-360 76434-240 76434-362 76434-242 76434-364 76434-244 76434-366 76434-400 76434-246 76434-368 76434-248 76434-402 76434-238 76434-190 76434-192 76434-194 76434-350 76434-196 76430-094 76434-230 76434-198 76434-352 76434-232 76430-096 76434-354 76434-234 76430-098 76434-356 76434-236 76434-358 76434-348 76434-228
Avantor® Hichrom HI-1, GC Columns
Chromatography Columns
Really versatile general purpose apolar phase for a wide range of applications, including (for example): PCBs, simulated distillation, gases, essential oils, hydrocarbons analysis, pesticides, semivolatiles, oxygenates. HI-1 is a crossbonded phase and assure a long lifetime even under high temperature limits.
  • Phase composition: Crossbonded 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
  • Long lifetime
Max temperature limits may change depending on the film thickness.