Edge® 48-929 Medium-Duty Cut Resistant Industrial Gloves, Ansell
Supplier: Ansell Healthcare
48929060 48929070 48929080 48929090 48929100 48929110
76424-298CS 1580.04 USD
76424-298 76424-300 76424-302 76424-304 76424-306 76424-308
Edge® 48-929 Medium-Duty Cut Resistant Industrial Gloves, Ansell
Gloves
Cost effective solution for medium weight applications requiring cut resistance, excellent grip, and strong oil-repellence.
  • Excellent abrasion resistance
  • Double layer nitrile coating keeps hands dry and comfortable when handling oily parts and tools
  • Elastic knitwrist ensures snug fit
  • Applicable in oily environments
Made of a seamless liner, fully dipped into NBR coating. The gloves are fully NBR dipped to protect skin from oil contamination. The palm rough NBR coating ensures a highly secure grip in oily environments. Use primarily for aerospace,More Product Information