5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na)

Supplier: RPI

Synonyms: 5-Bromo-4-chloro-1H-indol-3-yl phosphate sodiumX-phosphate disodium saltSodium 5-bromo-4-chloro-1H-indol-3-yl phosphate

B74000-0.1 B74000-1.0 B74000-5.0
76345-278EA 129.92 USD
76345-278 76345-282 76345-280
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na)
5-Bromo-4-chloro-3-indolyl phosphate disodium salt (BCIP-Na)

Colorimetric substrate for Alkaline phosphatase activity in blotting immunohistochemical and cytochemistry techniques.


Forms a purple insoluble precipitate when used in conjunction with NBT.


Caution: Caution: Research or further manufacturing use only, not for food or drug use.

Formula: C₈H₄BrClNNa₂O₄P
MW: 370.43 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00036757
CAS Number: 102185-33-1

Order Now

Specification Test Results

Appearance: Off white to cream coloured solid
Identification (IR): Consistent with assigned structure
Inorganic Phosphate: ≤ 0.2 %
Storage Condition: -10 - -20 oC, Desiccated dark.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR