Benzamidine hydrochloride ≥98.5%

Supplier: RPI

Synonyms: Benzimidamide hydrochloride, Benzamidinium chloride, Phenylamidine hydrochloride, Benzene-1-carboximidamide hydrochloride

B12000-100.0 B12000-25.0
76345-094EA 195.34 USD
76345-094 76345-096
Benzamidine hydrochloride ≥98.5%
Benzamidine hydrochloride

Competitive serine protease inhibitor which forms stable complexes with and blocks the active sites of enzymes.


Target enzymes include trypsin, hymotrypsin, kallikzein and plasmin.


Caution: Caution: Research or further manufacturing use only, not for food or drug use.

Formula: C₇H₉ClN₂
Melting Pt: 86…88 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00066285
CAS Number: 1670-14-0
EINECS: 216-795-4

Order Now

Specification Test Results

Appearance: White crystalline powder
Assay (HPLC): ≥ 98.5 %
Loss on Drying: ≤ 2 %
Residue on Ignition: ≤ 0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR