TotalStain Q, Fluorescent Total Protein Stain Kit, Azure Biosystems
Supplier: Azure Biosystems
AC2225 AC2227
76421-036EA 242.85 USD
76421-036 76421-038
TotalStain Q, Fluorescent Total Protein Stain Kit, Azure Biosystems
Stains Microbiology Stains
Accurate and quantitative fluorescent total protein stain to perform normalization of Western blots.
  • Fluorescent total protein stain for Western blot membranes
  • Sufficient amount for 15 mini-blots
  • Simple 20-minute dye-and-wash protocol
  • Wide linear dynamic range between 1 to 50 μg of protein load across lysates of various tissue types
  • Compatible with both chemiluminescent and near-infrared fluorescence
TotalStain Q only adds a short 20-minute step to Western blot protocol, between protein transfer and blocking buffer incubation. Western blots can be imaged on both camera-based and scanner instruments with green channel or Safe-Dyes channel. The linear range between 1 to 50 μg of lysate makes it ideal for normalization of low-expressed proteins that require high loads of protein to reach the sensitivity for the detection of these targets.
This is compatible with subsequent Western blot detection using chemiluminescence or Near-Infrared (NIR) fluorescence.
This solves the issue of housekeeping proteins that lack linearity and normalization accuracy at high protein loads.
Delivery information: Kit contains 1 bottle of staining solution, 1 bottle of washing solution and 15 units of imaging folders.