Ammonium chloride 99,5 ACS
Supplier: RPI
|
Warning
|
Synonyms:
Ammonium muriate, Salmiac
A20424-1000.0
A20424-10000.0
A20424-500.0
A20424-25000.0
76344-752EA
98.38
USD
76344-752
76344-754
76344-756
77510-166
Ammonium chloride 99,5 ACS
Ammonium chloride
Ammonium chloride, an inorganic compound, is a white crystalline salt, highly soluble in water.
Caution: Research or further manufacturing use only, not for food or drug use.
Formula:
NH₄Cl MW: 53.49 g/mol Melting Pt: 335 Storage Temperature: Ambient |
MDL Number:
MFCD00011420 CAS Number: 12125-02-9 EINECS: 235-186-4 Merck Index: 13,00510 |
|
Specification Test Results
Appearance | White granular powder |
Assay | ≥ 99.5 % |
Calcium (Ca) | ≤ 0.001 % |
Heavy Metals (as Pb) | ≤ 0.0005 % |
Insoluble Matter | ≤ 0.005 % |
Iron (Fe) | ≤ 0.0002 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...