Agarose ME, Molecular Biology Grade medium electroendosmosis (EEO)

Supplier: RPI

A20085-100.0 A20085-500.0
76344-684EA 249.06 USD
76344-684 76344-686
Agarose ME, Molecular Biology Grade medium electroendosmosis (EEO)
Agarose

For DNA/RNA fragments > 1000bp


  • Intermediate EEO

Melting Pt: 260 °C
Storage Temperature: Ambient
MDL Number: MFCD00081294
CAS Number: 9012-36-6
EINECS: 232-731-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=526c20a29610acf6900f898ff245c5602558c785&vid=ae34e4c76a4d7838d4dc9501397f6741b1b97d5e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to off white powder
EEO 0.16 - 0.19 -Mr
Gelling Temperature 34.5 - 37.5 °C
Gel Strength ≥ 1000 g/cm2
Moisture Content ≤ 10.0 %
Sulfate Content ≤ 0.20 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR