Accessories for Vivapore Solvent Absorption Concentrators
Supplier: Sartorius
Vivapore®
VPA005 VP-AST0001-C VPA002 VPA006 VPA002 VP-AST0001-C VPA007 VP-ARV0010-D VPA005 VP-ARV0010-D VPA007 VPA006
76408-858PK 39.52 USD
CA77581-124 76408-858 77581-124 CA77581-122 CA76408-864 CA76408-858 77581-122 76408-860 CA76408-862 76408-864 CA76408-860 76408-862
Accessories for Vivapore Solvent Absorption Concentrators
Concentrators
Stand with simple push-fit handling, each stand can hold up to four Vivapore units during use.
  • Accessories for use with Vivapore® solvent absorption concentrators
  • Stands hold up to four Vivapore® units during use
  • Expansion reservoirs for increased sample capacity or diafiltration
Compatible with Vivapore® 5 and 10 solvent absorption concentrators.