Scrub Brush, Micronova
Supplier: Micronova
BRWH-05
76415-982EA 50.79 USD
76415-982
Scrub Brush, Micronova
Brushes
This scrub brush features an 8", iron-shaped plastic block handle with a comfortable, curved grip that protects hands from metal edges.
  • Detail oriented scrub brush
  • Curved handle with stiff polyester bristles
  • Designed for tight crevices, rolled lips, and corners of equipment
  • Durable materials will not crack or splinter with use
The stiff, staple-set polyester bristles are set in a flat V-shaped pattern for maximum pan contact.
Certifications: FDA compliant base and bristles.