HPLC Hemp Analyzer, Shimadzu Scientific Instruments
Supplier: Shimadzu
220-91660-31
76415-476EA 80167.76 USD
76415-476
HPLC Hemp Analyzer, Shimadzu Scientific Instruments
Chromatography Systems Complete HPLC Systems
A complete turnkey HPLC analyzer designed for quantitative determination of cannabinoid content in hemp​.
  • Includes all required hardware, software, consumables, and analysis workflow
  • Run samples in the shortest possible time
The high sensitivity hemp analyzer is a fully applicated HPLC system based on the Shimadzu i-Series HPLC with built-in PDA detector for quantitative potency determination of hemp matrices.
High throughput method: When high sample throughput is paramount - 60 samples per 8 hr. day. Quantitative for ten cannabinoids (no THCV) in under eight minutes. Developed in collaboration with commercial testing laboratories.
High sensitivity method: When THCV is important, yet with good sample throughput - 48 samples per 8 hr. day. Quantitative for 11 cannabinoids (including THCV) in under ten minutes. The method provides the sharpest chromatographic peaks and best sensitivity.
High resolution method: For baseline separation of 11 plus components, including THCV. Minimum resolution is 2.0. 16 samples per 8 hr. day. This is the preferred method when performing research, adding additional compounds, or state regulations change.
Delivery information: The package includes all hardware, software, and consumables for installation and validation, plus analytical methods and batch files for running the three most common HPLC methods for analysis of cannabinoids. Including HPLC columns, certified standard mixture, solvents, computer and custom hemp software.