StrepTrap XT Prepacked Chromatography Columns, Cytiva
Supplier: Cytiva
StrepTrap™
29401323 29401320 29401317 29401322 29401322 29401317 29401320 29401323
76382-798EA 711.03 USD
76382-798 CA76382-796 76382-796 CA76382-802 76382-802 CA76382-800 76382-800 CA76382-798
StrepTrap XT Prepacked Chromatography Columns, Cytiva
Chromatography Columns
StrepTrap XP is a column packed with Strep-Tactin® XT sepharose resin to purify Strep-tag® II and Twin-Strep-tag® recombinant proteins.
  • Fast purifications with simple elution
  • Physiological conditions and mild elution preserve target protein activity
  • Fast and easy regeneration
  • Compatible with a wide range of detergents, denaturants, and other additives
  • Ready to use prepacked column
StrepTrap XP is a chromatography resin for the purification of recombinant proteins tagged with Strep-tag® II and Twin-Strep-tag®, which provides higher affinity, resulting in higher yields and lower loss of valuable target protein.