NanoXact Silica-Shelled Silver Nanospheres 1 mg/ml Silica shell (high stability), nanoComposix
Supplier: NANOCOMPOSIX, INC.
AGSH100-10M AGSH100-1M AGSH100-30M AGSH100-5M AGSH50-30M AGSH50-30M AGSH100-10M AGSH100-1M AGSH100-30M AGSH100-5M
103819-776EA 1960 USD
103819-776 103819-778 103819-780 103819-782 103819-784 77679-866 77679-854 77679-856 77679-858 77679-860
NanoXact Silica-Shelled Silver Nanospheres 1 mg/ml Silica shell (high stability), nanoComposix
Beads Microcarriers
Silver nanoparticles shelled with silica, versatile surface for a wide variety of applications
  • High quality nanoparticles, precisely engineered and highly characterized
  • Unagglomerated and monodisperse
  • High concentration gold (1 mg/ml gold concentration)
  • Silica shell offers robust and versatile surface for a wide variety of applications
Spherical silver nanoparticles, 1 mg/ml silver concentration. Silica shell offers high stability in a variety of solvents and a versatile surface for attaching functional groups or creating hydrophobic and hydrophilic surfaces. Silica shell allows easier deposition onto substrates and incororation into many different solvents, resins, and curable materials because the silica shell provents metal nanoparticles from touching directly / aggregating, helping to preserve the particles' nano and optical properties.
Each batch is extensively characterized to meet highest quality standards, ensuring reliable and reproducible results. All products supplied with batch-specific COA's with TEM, DLS, Zeta, and UV-Vis. to meet the highest quality standards
Colloidal silver has applications in Diagnostics, Antimicrobial coatings, Electronics and Photovoltaics, SERS, Optics, Nanotoxicology, Plasmonics (SPR / Surface Plasmon Resonance), novel Color Enhancements and Pigments, Molecular Detection, Sensors, and Photothermal and Microwave-based Therapies.
Custom formulations and ISO13485/cGMP compliant materials can be provided on request.
Delivery information: Ships via FedEx Priority Overnight (Domestic). Cold packaging included.
Due to temperature-sensitive nature of these materials, they will only be shipped during the week in order to prevent products from sitting in an uncontrolled temperature environment over the weekend.
Caution: Do not freeze. Store at 2 to 8 °C and away from light when not in use to maximize shelf life. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.