Health Smart® Oximeters
Supplier: HEALTH SMART SE
40-815-000 40-816-000 40-817-000
470326-232CS 4195.85 USD
470326-232 470326-234 470326-236
Health Smart® Oximeters
Blood Pressure Monitors
Measurements in just a few seconds.
  • Available in two versions
  • Readings in seconds
  • Single button operation
  • Displays pulse and Spo2
While not intended for diagnostic or treatment use, these pulse oximeters are a fast reliable way to take intermittent readings. Great for training purposes, and for taking measurements with exercise labs in the classroom. Standard models display pulse and SPO2 in a single color display. The deluxe uses a two color display and also includes a pulse wave form graph.
Caution: Not for diagnostic use, or use during exercise.