Go Direct® Weather Sensor
Supplier: Vernier
GDX-WTHR
470326-508EA 130.39 USD
470326-508
Go Direct® Weather Sensor
Weather Stations
Easily monitor a wide variety of environmental factors with just one sensor.
  • Rechargeable battery - Ready when you are
  • Wired or wireless - the flexibility to choose
  • Perfect for middle school or high school environmental science
Go Direct® weather is an affordable, wireless, handheld sensor used to measure ambient temperature, humidity, wind speed, wind chill, dew point, barometric pressure, and more. The robust rechargeable battery offers long battery life and provides always-ready operation when using the sensor wirelessly. Monitor battery life directly from free Graphical AnalysisTM 4 app or LabQuest 2 App. Should the battery run low, simply connect the sensor to the charging cable and keep on collecting - no interruptions or inconvenience to you or your students. This sensor also connect directly to your mobile device, ChrombookTM, or computer using their free Graphical AnalysisTM 4 app - no additional equipment or software purchases are necessary. They can be used wired via USB or wirelessly via Bluetooth® wireless technology, allowing you to choose the solution best for your classroom or laboratory.