Para-Ban Paraffin Cleaner/Repellent Spray, IMEB
Supplier: IMEB
Para-Ban
PB-604
103783-062CS 140 USD
103783-062
Para-Ban Paraffin Cleaner/Repellent Spray, IMEB
Cleaning Agents Detergents and Surface Cleansers
As the name implies, Para-Ban is a reliable paraffin repellent that will keep microtomes and adjacent surfaces free of paraffin. Apply with paper towel or gauze pad to desired surface.
  • Safe on any metal or plastic surface
  • Cleans and repels in one simply spray
  • Six 4 oz. bottles per case
  • Apply with paper towel or gauze pad
Simply spray Para-Ban on any surface where paraffin normally sticks, such as table tops and most microtomes. The spary is safe on any metal or plastic surface, and is used by histologists everywhere. Effectively cleans and repels in one simple spray.