Male Luer with 25G Connector, SAI Infusion Technologies
Supplier: SAI Infusion Technologies
ML-25
76408-384EA 15.83 USD
76408-384
Male Luer with 25G Connector, SAI Infusion Technologies
Fittings and Connectors
SAI offers a variety of connectors to simplify tubing and catheter access. These luered connectors can be used anywhere that catheter access is needed, such as harnesses, extension lines, catheters and Huber needles.
  • Male luer with attached pin
  • 25G stainless steel pin connector
  • Smooth, burr free 25G pin
  • Easy luer connection to 25G lines
SAI's luered connectors are compatible with any standard luer fitting, for a simple, quick and secure luer connection. These luered fittings offer unlimited flexibility for all of your research needs.