Paraffin, Tek-Select® IP (Infiltration) for embedding
Supplier: IMEB INC MS
Synonyms:
Wax Paraffin
, Paraffin wax
Tek-Select® IP (Infiltration) embedding paraffin is recommended for technicians who want to improve their infiltration processing and is excellent when used with microwave tissue processors.
- Improve infiltration processing
- Excellent when used with microwave tissue processors
- Melts uniformly at 55…57 °C
Tek-Select® IP (TS2IP) is a blend of highly refined paraffin with a lower concentration of polymers, which enhances infiltration and allows for proper fluid exchange. TS2IP melts uniformly at 55…57 °C. Water bath should be set between 38…42 °C. All Tek-Select paraffin products offer excellent ribboning of tissue sections, even at 2 - 3 microns. Choose Tek-Select® IP for improved infiltration during conventional or microwave processing.
Delivery information: Four 4.4 lb bags per case
Melting Pt:
55…57 °C |
CAS Number:
8002-74-2 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...