Tetrahydrofuran ≥99.7% unstabilised, AcroSeal™, Spectrophotometric Grade

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Oxolane, 1,4-epoxy-Butane, Butylene oxide, Butane alpha,delta-oxide, Tetramethylene oxide, THF, Oxacyclopentane, Cyclotetramethylene oxide, Diethylene oxide

Packaged under argon.

047329-AK 047329-K2 047329-M1
AA47329-AKEA 61.1 USD
AA47329-AK AA47329-K2 AA47329-M1
Tetrahydrofuran ≥99.7% unstabilised, AcroSeal™, Spectrophotometric Grade
Tetrahydrofuran

AcroSeal™ septum bottled solvent.


  • Supports optimum performance of your moisture-sensitive work

Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 66
Melting Pt: -108.5
Density: 0.889
Flash Pt: –17°C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR