EzyFit Twin Sample Tubes, PerkinElmer
Supplier: PerkinElmer
N0777462
10139-436PK 528.63 USD
10139-436
EzyFit Twin Sample Tubes, PerkinElmer
Tubing Transfer Tubing
These EzyFit Twin sample tubes are for use with MicroMist™ and SeaSpray™ nebulizers in Optima® 4x00/5x00/7x00/8x00 and Avio® 200/500 ICP-OES series instruments in addition to ELAN® 5000/6x00/9000/DRCs and NexION® 300/350/1000/2000 ICP-MS series instruments. It includes six sample tubes.
These sample tubes are replacement tubing for internal standard kits. It is included in the Non HF-Resistant internal standard kit (10139-144).