Forceps, Adson, Excelta Corp®
Supplier: Excelta
AF-2-SE AF-1-SE 10G-SA AF-4-SE AF-3-SE F-6-SE 10H-S-SE
75880-458EA 3.67 USD
75880-458 75880-724 10066-444 75880-460 75880-462 89411-852 75880-464
Forceps, Adson, Excelta Corp®
Forceps
This Adson dissection forceps is used for holding and manipulating delicate tissue.
  • Wide serrated thumb area for ergonomics
  • Excellent for handling delicate tissue
  • Cleanroom safe, stainless steel
This Adson stainless steel tissue forceps is used for holding and manipulating delicate tissues. These forceps have a wide, flat thumb grasp area that is commonly serrated. The serrated tips provide more 'sure' grip. The satin finish reduces glare.