Dura-Mag™ Stirrers, Magnetic, 5-Position, 230 V
Supplier: Chemglass
CLS-4100-A4E CLS-4100-A6E CLS-4100-04E CLS-4100-06E
76333-474EA 4166.99 USD
76333-474 76333-482 76333-472 76333-480
Dura-Mag™ Stirrers, Magnetic, 5-Position, 230 V
Stirrers Magnetic Stirrers, Multi-position
Digital, front-end, magnetic stirrers are designed for both cell culture in spinner flasks as well as general laboratory procedures requiring the use of magnetic stirring equipment.
  • 230 V model
Dura-Mag™ stirrers feature brushless, maintenance-free DC motors with drive assemblies designed for quiet operation. Stainless steel covers are stamped to show the vessel positions for accurate placement and optimum performance. Optional factory installed motion alarm available. Speed range: 25 to 400 rpm.
Caution: 230 V model.