Vernier Go Direct® Glass-Body pH Sensor
Supplier: VERNIER SOFTWARE & TECH SE
GDX-GPH
470320-004EA 158.99 USD
470320-004
Vernier Go Direct® Glass-Body pH Sensor
Electrodes pH Electrodes
Use the Go Direct Glass-body pH to measure pH in aqueous, heterogeneous, and organic solutions.
  • pH range of 0 to 14
  • Glass shaft, sealed combination electrode with Ag/AgCl reference
  • Shaft diameter of 12 mm outer.Ø
  • Shaft material is glass
This high-quality glass body pH sensor can be used in non-aqueous solutions and solutions that contain organic solvents, strong acids, or strong bases. The electrode features a sealed, gel-filled Ag-AgCl combination reference electrode and uses a BNC connector.
Go Direct sensors connect directly to your mobile device, Chromebook™, or computer using free Graphical Analysis™ 4 app - no additional equipment or software purchases are necessary. Go Direct sensors can be used wired via USB or wirelessly via Bluetooth® wireless technology, allowing you to choose the solution best for your classroom or laboratory.
Delivery information: It includes Go Direct Glass-body pH electrode BNC, Go Direct electrode amplifier, electrode tip guard, pH storage solution bottle with pH storage solution, Micro USB cable