Avantor® ACE® C8-300, HPLC Columns, 3 µm
Supplier: Avantor
ACE-212-0501 ACE-212-0502 ACE-212-0503 ACE-212-0504 ACE-212-0546 ACE-212-10005S ACE-212-1002 ACE-212-1003 ACE-212-1004 ACE-212-1046 ACE-212-0102GD ACE-212-0103GD ACE-212-1502 ACE-212-1546 ACE2121546MVK ACE-212-7502
76389-584EA 785.87 USD
76389-584 76389-586 76389-588 76389-590 76389-592 76389-594 76389-596 76389-598 76389-600 76389-602 76389-604 76389-606 76389-608 76389-610 76389-612 76389-614 76389-616 76389-618 76389-620 76389-622 76389-624 76389-626 76389-556 76389-558 76389-560 76389-562 76389-564 76389-566 76389-568 76389-570 76389-572 76389-574 76389-576 76389-578 76389-580 76389-582 76389-628 76389-630 76389-632 76389-634 76389-636 76389-638 76389-640 76389-642 76389-644 76389-646 76389-648 76389-650 76389-652 76389-654 76389-656 76389-658 76389-660 76389-662 76389-664 76389-666 76389-668 76389-670 76389-672 76389-674 76389-676 76389-678 76389-680 76389-682 76389-684 76389-686 76389-688 76389-690 76389-692 76389-694 76389-696 76389-698 76389-700 76389-702 76389-704 76389-706
Avantor® ACE® C8-300, HPLC Columns, 3 µm
Chromatography Columns
Avantor® ACE® C8-300 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (3, 5 and 10 µm) and complementary phases
  • Designed for peptide and protein separations
  • Method validation kits contain three different batches of the same phase - available on request
These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.