Avantor® ACE® C18-300, HPLC Columns, Microbore, 5 µm
Supplier: Avantor
ACE-221-0301 ACE-221-0501 ACE-221-1501
76390-296EA 1089.56 USD
76390-296 76390-298 76390-300 76390-302 76390-320 76390-170 76390-172 76390-174 76390-176 76390-186 76390-188 76390-190 76390-192 76390-214 76390-216 76390-218 76390-220 76390-238 76390-240 76390-242 76390-244 76390-266 76390-268 76390-270 76390-272 76390-324 76390-326 76390-338 76390-340 76390-342 76390-344
Avantor® ACE® C18-300, HPLC Columns, Microbore, 5 µm
Chromatography Columns
Avantor® ACE® C18-300 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative. These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.
  • Designed for peptide and protein separations
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle
More Product Information