Ward's® PCR Plates
Supplier: VWR International
470324-964 470324-966
470324-964BG 37.2 USD
470324-964 470324-966
Ward's® PCR Plates
Microplates PCR Plates
Choose these high quality PCR plates for your classroom experiments.
  • Alpha-numeric reference grids
  • Compatible with standard thermal cyclers
  • Compatible with ward's® pre-cut sealing film (available separately)
  • Available by the bag or by the case, buy only what you need
Designed to be compatible with most popular thermal cyclers, ward's® PCR plates are held to very tight dimensional tolerances with ultra-thin walls for optimal thermal transfer.
Certifications: These are for educational purposes, no certificates of quality are available.