CyQuanol Disinfectant Solutions, Contec®
Supplier: Contec
FBT32CQ SBT32CQ FBC128CQ SBC128CQ
76382-886CS 344.1 USD
76382-886 76382-888 76382-890 76382-892
CyQuanol Disinfectant Solutions, Contec®
Cleaning Agents Disinfectants
Blend of quaternary ammonium and ethyl alcohol.
  • 0.2 micron filtered for the most critical environments
  • Bactericidal, virucidal, fungicidal and tuberculocidal
  • 1-minute contact time makes it easier to achieve dwell time
  • No pre-clean required for this one-step cleaner disinfectant
  • Bleach-free makes it compatible with common cleanroom surfaces
It is a ready to use cleaner/disinfectant that generates low residue.
CyQuanol has a 1-minute bactericidal, virucidal, tuberculocidal and fungicidal contact time. It is bleach-free, making it an excellent choiceMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable