Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99,9 Dried, Ultra Pure Grade Molecular_Biology
Supplier: RPI
Synonyms:
2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris), Tromethamine, Tris(hydroxymethyl)methylamine, 2-Amino-2-(hydroxymethyl)propane-1,3-diol, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Trizma(R) base, TRIS base, THAM
T60040-100.0
T60040-1000.0
T60040-10000.0
T60040-250.0
T60040-25000.0
T60040-500.0
T60040-5000.0
T60040-200.0
76348-638EA
37.08
USD
76348-638
76348-640
76348-642
76348-644
76348-646
76348-648
76348-650
77510-474
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99,9 Dried, Ultra Pure Grade Molecular_Biology
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Highly purified for use in molecular biology applications.
Caution: Research or further manufacturing use only, not for food or drug use.
Formula:
NH₂C(CH₂OH)₃ MW: 121.14 g/mol Boiling Pt: 357 Melting Pt: 170…172 °C Storage Temperature: Room Temperature |
MDL Number:
MFCD00004679 CAS Number: 77-86-1 EINECS: 201-064-4 Merck Index: 13,09842 |
|
Specification Test Results
Appearance: | White Crystalline Powder |
Purity (Dried Basis By GC): | ≥ 99.9 % |
Related Substances By GC: | ≤ 0.1 % |
Water: | ≤ 0.2 % |
DNase, RNase, protease: | None Detected |
pH (5% in water): | 10.0 - 11.5 |
Heavy Metals (as Pb): | ≤ 5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...