MICROCHEM® AlphaTec® 600 Oversleeves, Ansell
Supplier: Ansell
Microchem® by AlphaTec®
GR40-W-92-600-00
76207-746CS 773.62 USD
76207-746
MICROCHEM® AlphaTec® 600 Oversleeves, Ansell
Disposable Garments
Model 600 oversleeves are designed for use with the MICROCHEM® by AlphaTec® 4000 coveralls.
  • Multi-layer non woven barrier laminate fabric
  • Elasticated at both ends
  • Welded seams
Packaging: Sleeves are packaged 100 pairs per case.