2-Propanol ACS

Supplier: Ricca Chemical
Danger

Synonyms: Isopropyl rubbing alcohol, sec-Propanol, Isopropanol, 2-Hydroxypropane, Isopropyl Rubbing Alcohol, 2-Hydroxy propane, IPA, sec-Propyl Alcohol, Dimethyl carbinol, Propane, 2-hydroxy-, Isopropyl alcohol, Propan-2-ol

RSOI0020-4C RSOI0020-1C RSOI0020-20F RSOI0020-4C1
RCRSOI0020-4CEA 227 USD
RCRSOI0020-4C RCRSOI0020-1C RCRSOI0020-20F RCRSOI0020-4C1 RCRSOI00204X4C
2-Propanol ACS
2-Propanol
Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82
Melting Pt: -89
Density: 0.7808
Flash Pt: 12°C
MDL Number: MFCD00011674
CAS Number: 67-63-0
UN: 1219
ADR: 3 II
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay min 99.5 %
Color (APHA) max 10
Solubility in Water To Pass Test
Carbonyl Compounds: Acetone max 0.002 %
Carbonyl Compounds: Propionaldehyde max 0.002 %
Water (H₂O) max 0.2 %
Residue after evaporation (Non-Volatile Matter) max 0.001 %
Titrable acid or base max 0.0001 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR