Labels 'Flammable Gas', Labelmaster
Supplier: Label Master
HML7S HML7C HML7
101378-994PK 11.35 USD
101378-994 101378-392 101376-396
Labels 'Flammable Gas', Labelmaster
Labels
Help take the guessing out of shipping label solutions with Worded Flammable Gas Labels, revised to meet the design requirements of 49 CFR 172.407.
  • Printed with light-fast inks for fade resistance
  • Adhesives formulated for use in various climates and environments
  • Strong holds on corrugated boxes
  • Once affixed, labels won't fade or fall off
  • Outstanding chemcial and abrasion resistance
In addition to complying with directives set by the DOT, these labels also meet the design and durability standards of 49 CFR, ICAO, and other international air regulations.