Labels 'Toxic Gas', Labelmaster
Supplier: Label Master
HML27 HML27S
101376-378PK 58.71 USD
101376-378 101378-976
Labels 'Toxic Gas', Labelmaster
Labels
Employ a fool-proof shipping label solution with Worded Toxic labels, revised to meet the design requirements of 49 CFR 172.407.
  • Printed with light-fast inks for fade resistance
  • Adhesives formulated for use in various climates and environments
  • Strong holds on corrugated boxes
  • Once affixed, labels will not fade or fall off
  • Outstanding chemical and abrasion resistance
In addition to complying with directives set by the DOT, these labels also meet the design and durability standards of 49 CFR, ICAO, and other international air regulations.