Kanamycin sulfate

Supplier: RPI
Danger

K22000-1.0 K22000-10.0 K22000-100.0 K22000-25.0 K22000-5.0
76346-390EA 44.52 USD
76346-390 76346-392 76346-394 76346-398 76346-400
Kanamycin sulfate
Kanamycin sulphate

Aminoglycoside antibiotic with a bactericidal action against most gram negative bacteria.


  • Soluble in water


Incorporates into sensitive bacterial cells by an active transport process. Binds to the 30S and to some extent the 50S subunits of the bacterial ribosome, inhibiting protein synthesis and generating errors in the transcription of the genetic code. Widely used as a selective agent for the incorporation of the NPT II (APH3') gene in plant tissue.


Caution: Research or further manufacturing use only, not for food or drug use.

Formula: C₁₈H₃₈N₄O₁₅S
Storage Temperature: Ambient
MDL Number: MFCD00070253
CAS Number: 25389-94-0

Order Now

Specification Test Results

Appearance White Powder
Chromatographic Purity Pass Test
Identification Pass Test
Loss on Drying ≤ 1.5 %
pH (1% Solution) 6.5 - 8.5
Potency ≥ 750 µg/mg
Residue on Ignition ≤ 1.0 %
Specific Rotation +116 - +123 °

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR