Dithiothreitol (DTT, Cleland's reagent) ≥98.0% (by iodometry), Molecular Biology Grade

Supplier: RPI
Warning

Synonyms: cis-2,3-Dihydroxy-1,4-butanedithiol, (R*,R*)-1,4-Dimercapto-2,3-butandiol, DTT, DL-1,4-Dithiothreitol

D11000-1.0 D11000-10.0 D11000-100.0 D11000-25.0 D11000-5.0 D11000-50.0
76345-718EA 32.69 USD
76345-718 76345-720 76345-722 76345-726 76345-728 76345-730 76345-732
Dithiothreitol (DTT, Cleland's reagent) ≥98.0% (by iodometry), Molecular Biology Grade
Dithiothreitol (DTT, Cleland's reagent)

Protective reagent for maintaining -SH groups in reduced state.


  • Soluble in water


DTT permeates the cell membrane protecting protein sulfhydryls thereby restoring enzyme activity lost by oxidation of the groups.


Caution: Research or further manufacturing use only, not for food or drug use.

Formula: C₄H₁₀O₂S₂
MW: 154.25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00004877
CAS Number: 3483-12-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5022472c3f3baef75a703791c548fc6504df810f&vid=81529732db015cb66c1f106092309ae5714325b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White free flowing powder,hygroscopic
Assay (By Iodimetry) ≥ 98.0 %
Identification Complies
Melting Range 37.0 - 44.0 °C
Related Substance (UV Spectroscopy- Content of Oxidised DTT) ≤ 0.4 %
UV Absorbance @ 280nm 0.1M ≤ 0.10

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR