Carbenicillin disodium salt ≥92.0% (by HPLC) USP
Supplier: RPI
|
Danger
|
Synonyms:
6-[(Carboxyphenylacetyl)-amino]-3,3-dimethyl-7-oxo-4-thia-1-aza-bicyclo[3.2.0]heptane-2-carboxylic Acid, 2Na
, α-Carboxybenzylpenicillin disodium salt
C46000-5.0
C46000-0.25
C46000-25.0
C46000-1.0
C46000-100.0
C46000-500.0
C46000-50.0
76345-522EA
783.45
USD
76345-522
76345-524
76345-520
76345-518
76345-526
77510-222
76345-516
Carbenicillin disodium salt ≥92.0% (by HPLC) USP
Carbenicillin disodium salt
USP Grade. Prevents bacterial cell wall synthesis. Inhibits crosslinking of peptidoglycan.
- Highly active against gram negative bacteria
- Commonly used for the elimination of Agrobacterium species after inoculation
- Sensitive to B-Lactamase
- Non-toxic to plant cells
- Plant cell culture tested
- Hygroscopic, soluble in water
Formula:
C₁₇H₁₆N₂O₆S·2Na MW: 422.363 g/mol Storage Temperature: Refrigerator |
CAS Number:
4800-94-6 EINECS: 225-360-8 |
Specification Test Results
Appearance | White to Off White Crystalline Powder |
Identification, Sodium Reaction | Pass Test |
Water | ≤ 6.0 % |
pH | 6.50 - 8.0 |
Clarity of Solution | Clear Solution |
Benzyl penicillin sodium | ≤ 5.0 % |
Chromatographic Purity (HPLC) | ≥ 92.0 % |
Assay (Anhydrous) | ≥ 88.0 % |
Bacterial Endotoxins | ≤ 0.05 EU/mg |
Iodine Absorbing Substances | ≤ 8.0 % |
Potency (Dried basis) | ≥ 770 µg/mg |
Grade | USP |
Country of Origin | Report |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...