DURAN® Premium Caps, DWK Life Sciences
Supplier: DWK Life Sciences
DURAN®
1088679 1129600 1088678
76263-496PK 49.88 USD
76263-496 76263-498 76263-738
DURAN® Premium Caps, DWK Life Sciences
Closures Bottle Closures
Cap is moulded from a pigment free polymer to reduce the risk of leaching.
  • Wide usable temperature range from –196 to +200 °C
  • Very high chemical resistance
  • Colorless PFA, with PTFE faced silicone cap liner
  • Autoclavable
Typical applications: Due to its exceptional properties, it is ideal for applications in the pharmaceutical industry, storage of aggressive materials, and demanding sterilisation processes such as hot air sterilisation or depyrogenation.