Aluminum Seals, Button-Top, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
73843A-13 73843D-13 73843A-20 73843D-20 73844B-13 73844C-13 73845A-20 73845B-20 73845C-20 73845D-20
66004-048CS 320.99 USD
66004-048 66004-052 66004-054 66004-056 66004-058 66004-060 66004-062 66004-064 66004-066 66004-068 66004-072 66004-074 66004-076 66004-078
Aluminum Seals, Button-Top, DWK Life Sciences
Closures Chromatography Vial Closures
Safe and easy-to-use button-top aluminum seals allow one-handed operation.
  • Autoclavable
  • Tamper evident
  • No sharp metal edges