Properties of Materials Kit
Supplier: United Scientific Supplies
PMTKIT-SET PMTKIT
470320-746EA 279.25 USD
470320-746 470320-744
Properties of Materials Kit
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
Fully investigate density, buoyancy and specific gravity.
  • Allows students to make a wide range of investigations with only a small number of items
  • Contains two sets of three metal cylinders - aluminum, copper, and steel
In one set the cylinders all have equal masses, and in the other they have equal volumes. The measuring instruments - cylinder, spring scale, and caliper - measure volume, mass, and length.
This combination of items allows for a wide range of basic investigations, addressing the concepts of mass, volume, density and specific gravity and extending to an exploration of buoyancy.
Delivery information: Includes manual.