Compound Bar Set
Supplier: United Scientific Supplies
BMSET04
470320-740EA 33.15 USD
470320-740
Compound Bar Set
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
Thermal expansion of different metals.
  • Four different metal combinations
  • Quick and easy tools for demonstrating thermal expansion
There are four bar combinations: Brass/Aluminum, Brass/Iron, Copper/Iron, and Brass/ Copper. Since each metal has a different coefficient of thermal expansion, they will create a unique curve when paired with a different metal.
Metal bars are stamped with the identification of each metal, Aluminum (Al), Iron (Fe), Copper (Cu). The Brass bar is not stamped.
Delivery information: Includes activity guide.