Control D-Dimer, Audit MicroControls, Inc.
Supplier: AUDIT Microcontrols
K072M-6
97068-948PK 268.86 USD
97068-948
Control D-Dimer, Audit MicroControls, Inc.
Controls and Calibration Verification Materials
D-Dimer control is an assayed, stable, lyophilized, bi-level reference control for use with assays designed to quantitate D-Dimer.
Three levels.