BD BBL™ Martin-Lewis Agar, Prepared Media Stacker™ Plates, BD Diagnostics
Supplier: BD Diagnostics
BD BBL™
221557 221557 221558
90001-388PK 327.67 USD
90001-388 CA90006-266 90006-266
BD BBL™ Martin-Lewis Agar, Prepared Media Stacker™ Plates, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Martin-Lewis agar is an enriched medium for the selective isolation of Neisseria species. Prepared plated media are designed for the isolation of microorganisms from samples or specimens and come in 100×15 mm petri-style configurations.
  • Ready for immediate use
These formulations developed for the selective isolation of pathogenic Neisseria, which is more inhibitory to gram-positive bacteria and yeasts than Thayer Martin agars. The concentration of vancomycin is increased from 3.0 to 4.0 µg/ml for greater inhibition of grampositive bacteria, and anisomycin is substituted for nystatin for improved inhibition of Candida albicans. This organism has been shown to inhibit Neisseria gonorrhoeae.